Pinacyanol chloride ≥93.0%

Leverantör: TCI EUROPE

Synonyms: 1,1'-Diethyl-2,2'-carbocyanine Chloride, Quinaldine Blue

P0156-1G P0156-100MG
TCIAP0156-1GEA 1300 SEK
TCIAP0156-1G TCIAP0156-100MG
Pinacyanol chloride ≥93.0%
Pinacyanol chloride
Formel: C₂₅H₂₅ClN₂
MW: 388,94 g/mol
MDL Number: MFCD00011974
CAS nummer: 2768-90-3
EINECS: 220-457-1
Merck Index: 14,08048

Order Now

Specifikation testresultat

Appearance Green to Dark green powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Argentmetric Titration) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR