Penta-O-acetyl-β-D-glucopyranose ≥99.0%

Leverantör: TCI EUROPE

Synonyms: β-D-Glucose Pentaacetate, Pentaacetyl-β-D-glucose, B-D-Glucose Pentaacetate, (2S,3R,4S,5R,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

P0028-100G P0028-500G
TCIAP0028-100GEA 775 SEK
TCIAP0028-100G TCIAP0028-500G
Penta-O-acetyl-β-D-glucopyranose ≥99.0%
Penta-O-acetyl-beta-D-glucopyranose
Formel: C₁₆H₂₂O₁₁
MW: 390,34 g/mol
MDL Number: MFCD00006597
CAS nummer: 604-69-3
EINECS: 210-074-8

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 99.0 %
Melting point 130.0 to 134.0 deg-C
Specific rotation [a]20/D +3.5 to +5.5 deg (C=5, CHCl3)
Solubility in hot Methanol almost transparency

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR