1-Octen-3-yl Acetate ≥98.0%

Leverantör: TCI
Varning

Synonyms: Acetic acid 1-Octen-3-yl ester

O0559-100ML O0559-25ML
TCIAO0559-100MLEA 1250 SEK
TCIAO0559-100ML TCIAO0559-25ML
1-Octen-3-yl Acetate ≥98.0%
1-Octen-3-yl Acetate
Formel: C₁₀H₁₈O₂
MW: 170.25 g/mol
Kokpunkt: 78°C
Flampunkt: 73°C
MDL Number: MFCD00036568
CAS nummer: 2442-10-6
EINECS: 219-474-7

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR