Sodium Pyridine-2-sulphinate ≥95.0%

Leverantör: TCI EUROPE
Varning

Synonyms: Sodium pyridine-2-sulphinate, Pyridine-2-sulphinic acid sodium salt

O0533-250MG O0533-1G
TCIAO0533-250MGEA 1300 SEK
TCIAO0533-250MG TCIAO0533-1G
Sodium Pyridine-2-sulphinate ≥95.0%
Sodium Pyridine-2-sulphinate
Formel: C₅H₄NNaO₂S
MW: 165,14 g/mol
MDL Number: MFCD20483664
CAS nummer: 24367-66-6

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 93.0 area%
Purity (Nonaqueous Titration) min. 95.0 %
Melting point 283.0 to 287.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR