Oxymetazoline hydrochloride ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Fara

Synonyms: SOxymetazoline hydrochloride, 6-tert-butyl-3-(4,5-dihydro-1H-imidazol-2-ylmethyl)-2,4-dimethylphenol hydrochloride, Afrin hydrochloride

O0520-25G O0520-5G
TCIAO0520-25GEA 4875 SEK
TCIAO0520-25G TCIAO0520-5G
Oxymetazoline hydrochloride ≥98.0% (by HPLC, titration analysis)
Oxymetazoline hydrochloride
Formel: C₁₆H₂₄N₂O·HCl
MW: 296,839 g/mol
Smältpunkt: 303°C
MDL Number: MFCD00058147
CAS nummer: 2315-02-8
EINECS: 219-015-0
UN: 2811
ADR: 6.1,I
Merck Index: 14,06966

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR