Octan-2-yl (2,4-Dichlorophenoxy)acetate ≥96.0%
Leverantör: TCI EUROPE
Synonyms:
2,4-D Meptyl ester, 3-(2,4-Dichlorophenoxy)octan-2-yl acetate, 2,4-D IOE, 2,4-D Octan-2-yl ester, (2,4-Dichlorophenoxy)acetic acid Octan-2-yl ester
O0518-100MG
O0518-1G
TCIAO0518-100MGEA
1450
SEK
TCIAO0518-100MG
TCIAO0518-1G
Octan-2-yl (2,4-Dichlorophenoxy)acetate ≥96.0%
Octan-2-yl (2,4-Dichlorophenoxy)acetate
Formel:
C₁₆H₂₂Cl₂O₃ MW: 333,25 g/mol |
CAS nummer:
1917-97-1 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...