Orbifloxacin ≥98.0%
Leverantör: TCI
Synonyms:
1-Cyclopropyl-5,6,8-trifluoro-1,4-dihydro-7-(cis-3,5-dimethyl-1-piperazinyl)-4-oxoquinoline-3-carboxylic acid, 1-Cyclopropyl-7-(cis-3,5-dimethylpiperazin-1-yl)-5,6,8-trifluoro-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
O0516-100MG
O0516-500MG
TCIAO0516-100MGEA
790
SEK
TCIAO0516-100MG
TCIAO0516-500MG
Orbifloxacin ≥98.0%
Orbifloxacin
Formel:
C₁₉H₂₀F₃N₃O₃ MW: 395.38 g/mol |
MDL Number:
MFCD00864858 CAS nummer: 113617-63-3 Merck Index: 14,06862 |
Specifikation testresultat
Appearance | White to Orange to Green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...