2-Octyn-1-ol ≥98.0%
Leverantör: TCI
O0493-5ML
O0493-25ML
TCIAO0493-5MLEA
525
SEK
TCIAO0493-5ML
TCIAO0493-25ML
2-Octyn-1-ol ≥98.0%
2-Octyn-1-ol
Formel:
C₈H₁₄O Kokpunkt: 78°C Smältpunkt: -18°C Flampunkt: 91°C |
MDL Number:
MFCD00039542 CAS nummer: 20739-58-6 |
Specifikation testresultat
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...