OTX015 ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
HY-15743, Birabresib, (S)-2-(4-(4-Chlorophenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepin-6-yl)-N-(4-hydroxyphenyl)acetamide, (6S)-4-(4-chlorophenyl)-N-(4-hydroxyphenyl)-2,3,9-trimethyl-6H-thieno[3,2-f][1,2,4]triazolo[4,3-a][1,4]diazepine-6-acetamide
O0482-10MG
O0482-50MG
TCIAO0482-10MGEA
1100
SEK
TCIAO0482-10MG
TCIAO0482-50MG
OTX015 ≥98.0%
OTX015
Formel:
C₂₅H₂₂ClN₅O₂S MW: 492 g/mol Smältpunkt: 220°C Storage Temperature: Frys |
MDL Number:
MFCD26960949 CAS nummer: 202590-98-5 |
Specifikation testresultat
Appearance | White to Light yellow to Light orange powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 13.50 to 15.00 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...