Oxytetracycline hydrochloride ≥95.0% (by HPLC, titration analysis)
Leverantör: TCI
|
Varning
|
O0475-5G
O0475-25G
TCIAO0475-5GEA
310
SEK
TCIAO0475-5G
TCIAO0475-25G
Oxytetracycline hydrochloride ≥95.0% (by HPLC, titration analysis)
Oxytetracycline hydrochloride
Formel:
C₂₂H₂₅ClN₂O₉ |
MDL Number:
MFCD00135815 CAS nummer: 2058-46-0 EINECS: 218-161-2 UN: 3077 ADR: 9,III Merck Index: 14,06976 |
|
Specifikation testresultat
Appearance | Light orange to Yellow to Green powder to crystal |
Purity (HPLC) | min. 95.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Specific rotation [a]20/D | -170 to -185 deg (C=1, 1mol/L HCl) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...