Oxetan-3-yl 4-methylbenzenesulphonate ≥98.0%

Leverantör: TCI EUROPE

Synonyms: p-Toluenesulphonic acid 3-Oxetanyl ester, 3-Oxetanyl p-Toluenesulphonate, 3-Oxetanyl Tosylate

O0473-5G O0473-25G
TCIAO0473-5GEA 885 SEK
TCIAO0473-5G TCIAO0473-25G
Oxetan-3-yl 4-methylbenzenesulphonate ≥98.0%
Oxetan-3-yl 4-methylbenzenesulphonate
Formel: C₁₀H₁₂O₄S
MW: 228,27 g/mol
MDL Number: MFCD08544401
CAS nummer: 26272-83-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5918684ab038b52113bedf04a43618ed241dcac&vid=c5918684ab038b52113bedf04a43618ed241dcac&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 89.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR