4-Cyano-4-phenylcyclohexanone ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
4-Oxo-1-phenylcyclohexanecarbonitrile, 4-Cyano-4-phenylcyclohexanone
O0470-5G
O0470-25G
TCIAO0470-5GEA
1125
SEK
TCIAO0470-5G
TCIAO0470-25G
4-Cyano-4-phenylcyclohexanone ≥98.0%
4-Cyano-4-phenylcyclohexanone
Formel:
C₁₃H₁₃NO MW: 199,24 g/mol |
MDL Number:
MFCD00044816 CAS nummer: 25115-74-6 EINECS: 246-631-7 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 118.0 to 122.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...