2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI

Synonyms: 2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride, 3-(2-Oxo-1,2-dihydro-4-quinolinyl)-DL-alanine hydrochloride

O0468-25G O0468-5G
TCIAO0468-25GEA 1925 SEK
TCIAO0468-25G TCIAO0468-5G
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride ≥98.0% (by HPLC, total nitrogen)
2-Amino-3-(2-oxo-1,2-dihydroquinolin-4-yl)propanoic acid hydrochloride
Formel: C₁₂H₁₃ClN₂O₃
MW: 268.70 g/mol
Smältpunkt: 225°C
Storage Temperature: Kylskåp
MDL Number: MFCD03839816
CAS nummer: 4876-14-6

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Specific rotation [a]20/D -1.0 to +1.0 deg (C=0.1, DMSO)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR