2-n-Octylbenzotriazole ≥98.0%
Leverantör: TCI EUROPE
Synonyms: 2-Octyl-2H-benzo[d][1,2,3]triazole
O0430-200MG O0430-1G
TCIAO0430-200MGEA 710 SEK
TCIAO0430-200MG TCIAO0430-1G
2-n-Octylbenzotriazole ≥98.0%
2-n-Octylbenzotriazole
Formel: C₁₄H₂₁N₃
MW: 231,34 g/mol
Kokpunkt: 110°C
MDL Number: MFCD00963848
CAS nummer: 112642-69-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff28071f0c1f4b2ddc124578e7cd21493d7ce7a0&vid=ff28071f0c1f4b2ddc124578e7cd21493d7ce7a0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %