2-Octyl-1-dodecanol ≥90.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 2-n-Octyl-1-dodecanol, 2-Octyldodecan-1-ol

O0429-25ML O0429-500ML
TCIAO0429-25MLEA 215 SEK
TCIAO0429-25ML TCIAO0429-500ML
2-Octyl-1-dodecanol ≥90.0%
2-Octyl-1-dodecanol
Formel: C₂₀H₄₂O
MW: 298,56 g/mol
Kokpunkt: 161°C
Flampunkt: 113°C
MDL Number: MFCD01310428
CAS nummer: 5333-42-6
EINECS: 226-242-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4714375cc28f1018da5510ab38d321667ac455ac&vid=4714375cc28f1018da5510ab38d321667ac455ac&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR