Oxetan-3-ol ≥96.0%
Leverantör: TCI EUROPE
Varning
Synonyms: 3-Hydroxyoxetane
O0409-1G O0409-5G
TCIAO0409-1GEA 495 SEK
TCIAO0409-1G TCIAO0409-5G
Oxetan-3-ol ≥96.0%
Oxetan-3-ol
Formel: C₃H₆O₂
MW: 74,08 g/mol
Kokpunkt: 50°C
Flampunkt: 87°C
Storage Temperature: Kylskåp
MDL Number: MFCD09056790
CAS nummer: 7748-36-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9c84d00378fdd04085b46088403655439c631901&vid=9c84d00378fdd04085b46088403655439c631901&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Light yellow clear liquid to slightly cloudy liquid
Purity (GC) min. 96.0 %