N-n-Octyl-3,4-thiophenedicarboximide ≥98.0%

Leverantör: TCI

O0394-200MG O0394-1G
TCIAO0394-200MGEA 630 SEK
TCIAO0394-200MG TCIAO0394-1G
N-n-Octyl-3,4-thiophenedicarboximide ≥98.0%
N-n-Octyl-3,4-thiophenedicarboximide
Formel: C₁₄H₁₉NO₂S
MDL Number: MFCD23703118
CAS nummer: 773881-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=33fa135bea8f397dd6aeec1fb3e3cc78d74e8f62&vid=33fa135bea8f397dd6aeec1fb3e3cc78d74e8f62&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 122.0 to 126.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR