3,4'-Oxydiphthalic anhydride ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Fara

O0384-1G O0384-5G O0384-25G
TCIAO0384-1GEA 255 SEK
TCIAO0384-1G TCIAO0384-5G TCIAO0384-25G
3,4'-Oxydiphthalic anhydride ≥98.0% (by GC, titration analysis)
3,4'-Oxydiphthalic anhydride
Formel: C₁₆H₆O₇
MW: 310,22 g/mol
Storage Temperature: Frys
CAS nummer: 50662-95-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7263f64defe5f38ce354f531bf889cb038822c10&vid=7263f64defe5f38ce354f531bf889cb038822c10&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Methanolysis Method) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR