(2S,3aS,7aS)-2-Carboxyoctahydroindole ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: (2S,3aS,7aS)-Octahydro-1H-indole-2-carboxylic acid, H-Oic-OH

O0370-1G O0370-5G
TCIAO0370-1GEA 525 SEK
TCIAO0370-1G TCIAO0370-5G
(2S,3aS,7aS)-2-Carboxyoctahydroindole ≥98.0% (by GC, titration analysis)
(2S,3aS,7aS)-2-Carboxyoctahydroindole
Formel: C₉H₁₅NO₂
MW: 169,22 g/mol
Smältpunkt: 270°C
MDL Number: MFCD07782125
CAS nummer: 80875-98-5

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Specific rotation [a]20/D -46.0 to -50.0 deg (C=1, mathanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR