Oxcarbazepine ≥98.0% (by HPLC, total nitrogen)

Leverantör: TCI EUROPE

Synonyms: 10-Oxo-10,11-dihydro-5H-dibenzo[b,f]azepine-5-carboxamide, 10,11-Dihydro-10-oxo-5H-dibenzo[b,f]azepine-5-carboxamide

O0363-25G O0363-5G
TCIAO0363-25GEA 2050 SEK
TCIAO0363-25G TCIAO0363-5G
Oxcarbazepine ≥98.0% (by HPLC, total nitrogen)
Oxcarbazepine
Formel: C₁₅H₁₂N₂O₂
MW: 252,27 g/mol
Smältpunkt: 216°C
MDL Number: MFCD00865307
CAS nummer: 28721-07-5
EINECS: 249-188-8
Merck Index: 14,06929
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=682517710e0d473e494f35752448af78aa7fe455&vid=682517710e0d473e494f35752448af78aa7fe455&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR