N-n-Octyl-D-glucamine ≥98.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
Synonyms:
(2R,3R,4R,5S)-6-(Octylamino)hexane-1,2,3,4,5-pentaol, 1-Deoxy-1-(n-octylamino)-D-glucitol
O0358-25G
O0358-5G
TCIAO0358-25GEA
805
SEK
TCIAO0358-25G
TCIAO0358-5G
N-n-Octyl-D-glucamine ≥98.0% (by HPLC, titration analysis)
N-n-Octyl-D-glucamine
Formel:
C₁₄H₃₁NO₅ MW: 293,40 g/mol |
MDL Number:
MFCD00134352 CAS nummer: 23323-37-7 EINECS: 245-582-9 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 98.0 % |
Specific rotation [a]20/D | -16.0 to -22.0 deg (C=0.5, Methanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...