2-Oxo-1-imidazolidinecarbonyl chloride ≥97.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
1-Chlorocarbonyl-2-imidazolidinone, 2-Oxoimidazolidine-1-carbonyl chloride, 1-Chloroformyl-2-imidazolidinone
O0352-25G
O0352-5G
TCIAO0352-25GEA
3450
SEK
TCIAO0352-25G
TCIAO0352-5G
2-Oxo-1-imidazolidinecarbonyl chloride ≥97.0%
2-Oxo-1-imidazolidinecarbonyl chloride
Formel:
C₄H₅ClN₂O₂ MW: 148,55 g/mol Smältpunkt: 157°C Storage Temperature: Frys |
MDL Number:
MFCD02683529 CAS nummer: 13214-53-4 EINECS: 236-185-1 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (Argentmetric Titration) | min. 97.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...