Ethyl-3-oxoheptanoate ≥95.0%
Leverantör: TCI
Synonyms:
3-Oxoenanthic Acid Ethyl Ester, Ethyl 3-oxoheptanoate, 3-Oxoheptanoic Acid Ethyl Ester, Ethyl 3-Oxoenanthate, 3-Ketoheptanoic Acid Ethyl Ester
O0216-10G
O0216-25G
TCIAO0216-10GEA
1525
SEK
TCIAO0216-10G
TCIAO0216-25G
Ethyl-3-oxoheptanoate ≥95.0%
Ethyl-3-oxoheptanoate
Formel:
C₉H₁₆O₃ Kokpunkt: 111°C |
MDL Number:
MFCD00143188 CAS nummer: 7737-62-4 |
|
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...