4-Octylaniline ≥97.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 4-n-Octylaniline

O0186-25G O0186-5G
TCIAO0186-25GEA 2575 SEK
TCIAO0186-25G TCIAO0186-5G
4-Octylaniline ≥97.0% (by GC, titration analysis)
4-Octylaniline
Formel: C₁₄H₂₃N
MW: 205,34 g/mol
Kokpunkt: 311°C
Storage Temperature: Kylskåp
MDL Number: MFCD00007929
CAS nummer: 16245-79-7
EINECS: 240-358-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b23fe2eba7e45020c5cc5dd0e373814d542502df&vid=b23fe2eba7e45020c5cc5dd0e373814d542502df&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White or Colorles to Yellow to Orange powder to lump to clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR