2-Octenylsuccinic anhydride (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
Leverantör: TCI
|
Varning
|
O0040-500G
O0040-25G
TCIAO0040-500GEA
3475
SEK
TCIAO0040-500G
TCIAO0040-25G
2-Octenylsuccinic anhydride (cis and trans mixture) cis- and trans-mixture ≥95.0% (by GC, titration analysis)
2-Octenylsuccinic anhydride (cis and trans mixture)
Formel:
C₁₂H₁₈O₃ Flampunkt: 147°C |
MDL Number:
MFCD00047133 CAS nummer: 42482-06-4 EINECS: 247-899-8 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % (total of isomers) |
Purity (Morpholine Method) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...