Octadecylsuccinic anhydride ≥95.0%

Leverantör: TCI
Varning

O0018-25G O0018-250G
TCIAO0018-25GEA 815 SEK
TCIAO0018-25G TCIAO0018-250G
Octadecylsuccinic anhydride ≥95.0%
Octadecylsuccinic anhydride
Formel: C₂₂H₄₀O₃
MDL Number: MFCD00014551
CAS nummer: 47458-32-2
EINECS: 256-316-6

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (Morpholine Method) min. 95.0 %
Melting point 77.0 to 81.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR