Octadecylamine acetate >60.0%

Leverantör: TCI
Varning

Synonyms: Stearylamine Acetate, Aminooctadecane Acetate

O0015-25G O0015-500G
TCIAO0015-25GEA 230 SEK
TCIAO0015-25G TCIAO0015-500G
Octadecylamine acetate >60.0%
Octadecylamine acetate
Formel: C₁₈H₃₉N
MW: 329.57 g/mol
MDL Number: MFCD00060198
CAS nummer: 2190-04-7
EINECS: 218-583-7

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR