Phenyl stearate ≥98.0%

Leverantör: TCI

Synonyms: Stearic Acid Phenyl Ester

O0007-25G O0007-5G
TCIAO0007-25GEA 5450 SEK
TCIAO0007-25G TCIAO0007-5G
Phenyl stearate ≥98.0%
Phenyl stearate
Formel: C₂₄H₄₀O₂
Kokpunkt: 267°C
MDL Number: MFCD00026663
CAS nummer: 637-55-8
EINECS: 211-291-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=62178a1556aa30faf37033a13b427ed0478e76c9&vid=62178a1556aa30faf37033a13b427ed0478e76c9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Melting point 52.0 to 56.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR