1-Ethynyl-3-nitrobenzene ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
3-Nitrophenylacetylene, 1-Ethynyl-3-nitrobenzene, (3-Nitrophenyl)ethyne, (3-Nitrophenyl)acetylene
N1148-200MG
N1148-1G
TCIAN1148-200MGEA
645
SEK
TCIAN1148-200MG
TCIAN1148-1G
1-Ethynyl-3-nitrobenzene ≥98.0%
1-Ethynyl-3-nitrobenzene
Formel:
C₈H₅NO₂ MW: 147,13 g/mol Kokpunkt: 120°C |
MDL Number:
MFCD02181150 CAS nummer: 3034-94-4 EINECS: 221-232-0 |
Specifikation testresultat
Appearance | White to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...