9-(2-Naphthyl)anthracene ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
9-(Naphthalen-2-yl)anthracene
N1130-1G
N1130-5G
TCIAN1130-1GEA
990
SEK
TCIAN1130-1G
TCIAN1130-5G
9-(2-Naphthyl)anthracene ≥98.0%
9-(2-Naphthyl)anthracene
Formel:
C₂₄H₁₆ MW: 304,39 g/mol |
MDL Number:
MFCD18205835 CAS nummer: 7424-72-8 |
Specifikation testresultat
Appearance | White to Light yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 203.0 to 207.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...