Methyl oleate 96%
Leverantör: Thermo Fisher Scientific
H31358.14 H31358.22
H31358.14EA 3500 SEK
H31358.14 H31358.22
Methyl oleate 96%
Methyl oleate
Purity: 96% CAS No.: 112-62-9 Molecular Formula: C19H36O2 Formula Weight: 296.50 Flash Point: 163°(325°F) Refractive Index: 1.4520 Boiling Point: 168-170°/2mm Beilstein Reference: 1727037 Merck Reference: 14,6828 MDL No.: MFCD00009578
Formel: C₁₉H₃₆O₂
Kokpunkt: 168…170 °C (2 mmHg)
Smältpunkt: –20 °C
Densitet: 0,876
Flampunkt: 163 °C (325 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00009578
CAS nummer: 112-62-9
EINECS: 203-992-5
Merck Index: 14,6828