2-Nonyn-1-ol ≥98.0%

Leverantör: TCI
Varning

N1115-25ML N1115-5ML
TCIAN1115-25MLEA 2475 SEK
TCIAN1115-25ML TCIAN1115-5ML
2-Nonyn-1-ol ≥98.0%
2-Nonyn-1-ol
Formel: C₉H₁₆O
Kokpunkt: 111°C
Smältpunkt: –16°C
MDL Number: MFCD00041566
CAS nummer: 5921-73-3
EINECS: 227-646-8
UN: 1987
ADR: 3,III

Order Now

Specifikation testresultat

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR