cis-4-Nitrostilbene ≥97.0%

Leverantör: TCI EUROPE

Synonyms: (Z)-4-Nitrostilbene

N1085-200MG N1085-1G
TCIAN1085-200MGEA 1450 SEK
TCIAN1085-200MG TCIAN1085-1G
cis-4-Nitrostilbene ≥97.0%
cis-4-Nitrostilbene
Formel: C₁₄H₁₁NO₂
MW: 225,25 g/mol
CAS nummer: 6624-53-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5fa7e448394a137551f64a207d831e58fdc5adaa&vid=5fa7e448394a137551f64a207d831e58fdc5adaa&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light yellow to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Melting point 61.0 to 65.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR