1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
alpha-Methyl-6-nitropiperonyl alcohol, 1-(6-Nitrobenzo[d][1,3]dioxol-5-yl)ethanol, alpha-Methyl-6-nitro-1,3-benzodioxole-5-methanol
N1069-1G
N1069-5G
TCIAN1069-1GEA
1575
SEK
TCIAN1069-1G
TCIAN1069-5G
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol ≥98.0%
1-(6-Nitro-1,3-benzodioxol-5-yl)ethanol
Formel:
C₉H₉NO₅ MW: 211,17 g/mol |
MDL Number:
MFCD08274663 CAS nummer: 159873-64-0 |
Specifikation testresultat
Purity (GC) | min. 98.0 % |
Melting point | 86.0 to 90.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...