N-[4-(1-Naphthyl)phenyl]-4-biphenylamine ≥98.0%
Leverantör: TCI EUROPE
Varning
Synonyms: N-[4-(Naphthalen-1-yl)phenyl]-[1,1'-biphenyl]-4-amine, N-(4-(Naphthalen-1-yl)phenyl)-[1,1'-biphenyl]-4-amine
N1053-200MG N1053-1G
TCIAN1053-200MGEA 1575 SEK
TCIAN1053-200MG TCIAN1053-1G
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine ≥98.0%
N-[4-(1-Naphthyl)phenyl]-4-biphenylamine
Formel: C₂₈H₂₁N
MW: 371,48 g/mol
MDL Number: MFCD28138088
CAS nummer: 897921-59-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3b9dbea97b4f9e8f199cd1bfbeb166b200e7f33d&vid=3b9dbea97b4f9e8f199cd1bfbeb166b200e7f33d&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 99.0 to 103.0 deg-C