2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Perfluorobutyl-1,2-epoxypropane

N1041-5G N1041-25G N1041-100G
TCIAN1041-5GEA 790 SEK
TCIAN1041-5G TCIAN1041-25G TCIAN1041-100G
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane ≥98.0%
2,2,3,3,4,4,5,5,5-Nonafluoropentyloxirane
Formel: C₇H₅F₉O
MW: 276,10 g/mol
Kokpunkt: 132°C
MDL Number: MFCD00236085
CAS nummer: 81190-28-5
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1eb9b981061d21a6ddcaca9915f352354f505fb0&vid=1eb9b981061d21a6ddcaca9915f352354f505fb0&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR