Nitazoxanide ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: NTZ

N1031-200MG N1031-1G N1031-5G
TCIAN1031-200MGEA 525 SEK
TCIAN1031-200MG TCIAN1031-1G TCIAN1031-5G
Nitazoxanide ≥98.0% (by HPLC, titration analysis)
Nitazoxanide
Formel: C₁₂H₉N₃O₅S
MW: 307,28 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00416599
CAS nummer: 55981-09-4
EINECS: 259-931-8
Merck Index: 14,06567
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f86a77241736b2597290c617efa1448f48ff850&vid=3f86a77241736b2597290c617efa1448f48ff850&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 198.0 to 202.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR