2-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Leverantör: TCI EUROPE
Varning

Synonyms: 2-Nitrophenylboronic acid Pinacol ester, 2-Nitrobenzeneboronic acid pinacol ester, 2-Nitrophenylboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(2-nitrophenyl)-1,3,2-dioxaborolane, 1-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzene

N1006-1G N1006-5G
TCIAN1006-1GEA 455 SEK
TCIAN1006-1G TCIAN1006-5G
2-Nitrophenylboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
2-Nitrophenylboronic acid pinacol ester
Formel: C₁₂H₁₆BNO₄
MW: 249,07 g/mol
MDL Number: MFCD02179447
CAS nummer: 190788-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e28076f2e2e75c01e4d95a98562ad8fbec880482&vid=e28076f2e2e75c01e4d95a98562ad8fbec880482&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 45.0 to 49.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR