N-(1-Naphthyl)formamide ≥98.0%

Leverantör: TCI EUROPE

N1001-5G N1001-25G
TCIAN1001-5GEA 990 SEK
TCIAN1001-5G TCIAN1001-25G
N-(1-Naphthyl)formamide ≥98.0%
N-(1-Naphthyl)formamide
Formel: C₁₁H₉NO
MW: 171,20 g/mol
MDL Number: MFCD00086455
CAS nummer: 6330-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=505bd91771915de5b09e804046fcab46dd06310f&vid=505bd91771915de5b09e804046fcab46dd06310f&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 138.0 to 142.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR