Nefiracetam ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: N-(2,6-Dimethylphenyl)-2-(2-oxopyrrolidin-1-yl)acetamide

N0996-200MG N0996-1G
TCIAN0996-200MGEA 1775 SEK
TCIAN0996-200MG TCIAN0996-1G
Nefiracetam ≥98.0%
Nefiracetam
Formel: C₁₄H₁₈N₂O₂
MW: 246,31 g/mol
MDL Number: MFCD00209882
CAS nummer: 77191-36-7
Merck Index: 14,06439
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1ccf145b14995aaff8b72482a6054e257cd8bd1a&vid=1ccf145b14995aaff8b72482a6054e257cd8bd1a&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 149.0 to 153.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR