5-Norbornene-2-methanol ≥98.0%
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
Bicyclo[2.2.1]hept-5-ene-2-methanol, 5-Norbornen-2-methanol (mixture of isomers), 2-Hydroxymethyl-5-norbornene, Bicyclo[2.2.1]hept-5-en-2-ylmethanol
N0979-5G
N0979-25G
TCIAN0979-5GEA
325
SEK
TCIAN0979-5G
TCIAN0979-25G
5-Norbornene-2-methanol ≥98.0%
5-Norbornene-2-methanol
Formel:
C₈H₁₂O MW: 124,183 g/mol Kokpunkt: 120°C Flampunkt: 87°C |
MDL Number:
MFCD00167571 CAS nummer: 95-12-5 EINECS: 202-392-0 |
Specifikation testresultat
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 98.0 % (total of isomers) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...