4-Nitrophenyl-N-benzylcarbamate ≥98.0%

Leverantör: TCI

N0978-200MG N0978-1G
TCIAN0978-200MGEA 405 SEK
TCIAN0978-200MG TCIAN0978-1G
4-Nitrophenyl-N-benzylcarbamate ≥98.0%
4-Nitrophenyl-N-benzylcarbamate
Formel: C₁₄H₁₂N₂O₄
MDL Number: MFCD06796634
CAS nummer: 124068-97-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cd8e949a65225ced509960ac9da6e58d9eb726c9&vid=cd8e949a65225ced509960ac9da6e58d9eb726c9&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 95.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 130.0 to 134.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR