1-(4-Nitrophenyl)-2-piperidone ≥98.0%
Leverantör: TCI EUROPE
Synonyms:
1-(4-Nitrophenyl)piperidin-2-one
N0975-1G
N0975-5G
TCIAN0975-1GEA
525
SEK
TCIAN0975-1G
TCIAN0975-5G
1-(4-Nitrophenyl)-2-piperidone ≥98.0%
1-(4-Nitrophenyl)-2-piperidone
Formel:
C₁₁H₁₂N₂O₃ MW: 220,23 g/mol |
MDL Number:
MFCD04037154 CAS nummer: 38560-30-4 |
Specifikation testresultat
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 97.0 to 101.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...