3-Nitrochalcone ≥98.0%

Leverantör: TCI EUROPE
Varning

Synonyms: 3-Nitrobenzylidenacetophenone

N0936-1G N0936-5G
TCIAN0936-1GEA 630 SEK
TCIAN0936-1G TCIAN0936-5G
3-Nitrochalcone ≥98.0%
3-Nitrochalcone
Formel: C₁₅H₁₁NO₃
MW: 253,26 g/mol
MDL Number: MFCD00024571
CAS nummer: 614-48-2
EINECS: 210-384-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25aa72924b6c5db7b88fa4ca11c86ec32c49de25&vid=25aa72924b6c5db7b88fa4ca11c86ec32c49de25&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 142.0 to 146.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR