Nizatidine ≥97.0% (by HPLC, titration analysis)
Leverantör: TCI EUROPE
|
Varning
|
Synonyms:
N-[2-[[[2-[(Dimethylamino)methyl]-4-thiazolyl]methyl]thio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine, N-(2-(((2-((Dimethylamino)methyl)thiazol-4-yl)methyl)thio)ethyl)-N-methyl-2-nitroethene-1,1-diamine
N0933-5G
N0933-25G
TCIAN0933-5GEA
655
SEK
TCIAN0933-5G
TCIAN0933-25G
Nizatidine ≥97.0% (by HPLC, titration analysis)
Nizatidine
Formel:
C₁₂H₂₁N₅O₂S₂ MW: 331,45 g/mol |
MDL Number:
MFCD00865660 CAS nummer: 76963-41-2 Merck Index: 14,06660 |
Specifikation testresultat
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 97.0 area% |
Purity (Nonaqueous Titration) | min. 97.0 % |
Melting point | 130.0 to 134.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...