Nadifloxacin ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI EUROPE

Synonyms: 9-Fluoro-8-(4-hydroxypiperidin-1-yl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic acid, 9-Fluoro-8-(4-hydroxy-1-piperidinyl)-5-methyl-1-oxo-1,5,6,7-tetrahydropyrido[3,2,1-ij]quinoline-2-carboxylic Acid

N0931-200MG N0931-1G
TCIAN0931-200MGEA 645 SEK
TCIAN0931-200MG TCIAN0931-1G
Nadifloxacin ≥98.0% (by HPLC, titration analysis)
Nadifloxacin
Formel: C₁₉H₂₁FN₂O₄
MW: 360,39 g/mol
Storage Temperature: Kylskåp
MDL Number: MFCD00865081
CAS nummer: 124858-35-1
Merck Index: 14,06345
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78a7a748d99f426cf8dc4471b5bcff89f170c241&vid=78a7a748d99f426cf8dc4471b5bcff89f170c241&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR