5-Nitropicolinic acid ≥98.0% (by HPLC, titration analysis)

Leverantör: TCI
Varning

Synonyms: 5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid

N0930-1G N0930-5G
TCIAN0930-1GEA 855 SEK
TCIAN0930-1G TCIAN0930-5G
5-Nitropicolinic acid ≥98.0% (by HPLC, titration analysis)
5-Nitropicolinic acid
Formel: C₆H₄N₂O₄
MDL Number: MFCD04114182
CAS nummer: 30651-24-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=91359c99cc8d11f59488621477e8a1cbdc58d384&vid=91359c99cc8d11f59488621477e8a1cbdc58d384&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR