trans,cis-2,6-Nonadienal ≥95.0%
Leverantör: TCI EUROPE
|
Varning
|
N0836-5G
N0836-1G
TCIAN0836-5GEA
2100
SEK
TCIAN0836-5G
TCIAN0836-1G
trans,cis-2,6-Nonadienal ≥95.0%
trans,cis-2,6-Nonadienal
Formel:
C₉H₁₄O MW: 138,21 g/mol Kokpunkt: 90°C Flampunkt: 83°C |
MDL Number:
MFCD00007009 CAS nummer: 557-48-2 EINECS: 209-178-6 |
Specifikation testresultat
Appearance | Colorless to Light orange to Yellow clear liquid |
Purity (GC) | min. 95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...