5-(4-Nitrophenyl)-2-furaldehyde ≥98.0%
Leverantör: TCI
|
Varning
|
N0820-25G
N0820-5G
TCIAN0820-25GEA
1425
SEK
TCIAN0820-25G
TCIAN0820-5G
5-(4-Nitrophenyl)-2-furaldehyde ≥98.0%
5-(4-Nitrophenyl)-2-furaldehyde
Formel:
C₁₁H₇NO₄ |
MDL Number:
MFCD00124191 CAS nummer: 7147-77-5 EINECS: 230-459-4 |
Specifikation testresultat
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 205.0 to 209.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...