Neopentyl glycol diacrylate ≥89.0% stabilised
Leverantör: TCI EUROPE
Fara
Synonyms: 2,2-Dimethyl-1,3-propanediol Diacrylate, 1,3-Bis(acryloyloxy)-2,2-dimethylpropane
Stabilization: Stabilised with 4-methoxyphenol (MEHQ)
N0790-25G N0790-500G N0790-100G
TCIAN0790-25GEA 335 SEK
TCIAN0790-25G TCIAN0790-500G TCIAN0790-100G
Neopentyl glycol diacrylate ≥89.0% stabilised
Neopentyl glycol diacrylate
Formel: C₁₁H₁₆O₄
MW: 212,25 g/mol
Kokpunkt: 96°C
Smältpunkt: 6°C
Flampunkt: 115°C
MDL Number: MFCD00048148
CAS nummer: 2223-82-7
EINECS: 218-741-5
UN: 2810
ADR: 6.1,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8e468f9e1532ffb9e87a75edf5df256a94aa57a0&vid=8e468f9e1532ffb9e87a75edf5df256a94aa57a0&prid=web&clid=SAPDAM
Order Now
Specifikation testresultat
Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 89.0 %